Wallcur® PRACTI-Ampule Safety Breakers
Supplier: WALLCUR, LLC.
905SB 907SB
470233-084BX 57.19 CAD
470233-084 470233-086 470233-090
Wallcur® PRACTI-Ampule Safety Breakers
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Safe And Disposable
  • Contains No Medication
  • Available In 2 Sizes
  • Designed For Safe And Accurate Opening Of Ampules
  • After Use, Dispose Of Broken Ampule And Breaker
Increase student confidence and competency during ampule handling and practice with these easy to use ampule breakers. Students will avoid direct finger contact with broken glass during and after practice thus eliminating the fear of cuts from poor technique. These safe, reliable and economical Ampule Breakers make ampule manipulation much easier and ampule breakage at the neck quicker and cleaner. Dispose of ampule and breaker together in a proper sharps container following practice. Available in 2 sizes to fit the WALLCUR PRACTI-Ampules.
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use