Wallcur® PRACTI-Naloxone
Supplier: WALLCUR, LLC.
1517NA 438NX
470232-740EA 34.19 CAD
470232-740 470232-868
Wallcur® PRACTI-Naloxone
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Increase Your Students' Naloxone Administration Knowledge!
  • Contains No Medication
  • Available In Either A Vial, Or Prefilled Syringe
  • Contents Are Safe For Use With Most Simulators
PRACTI-Naloxone HCI™ for clinical training. Provide your students with the most realistic and authentic practice necessary to master the skill of naloxone hydrochloride drug calculation and administration. In addition to being one of the most difficult-to-handle vial sizes, your students are now able to practice a variety of clinical scenarios related to opioid‐induced respiratory depression and overdose. Available in simulated vials, and prefilled syringes with nasal atomizer. A prefilled Code syringe is available with the PRACTI-Code Meds.
PRACTI-Naloxone HCI™ 0.4 mg/mL comes in a clear 1 mL vial with distilled water, allowing for safe practice with manikins and other injection simulators. PRACTI-Naloxone HCI™ is the perfect addition to your EMS, EMT, or Paramedic program.
PRACTI-Intranasal Naloxone™ Training Pack contains a (1 mg/mL) 2 mg per 2 mL pre-filled luer lock syringe and nasal atomizer device for clinical training only. Wallcur's PRACTI-Naloxone simulates the opioid antagonist drug, Naloxone for emergency opioid overdose. Teach a variety of skills to your students of nursing and EMT programs.
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use