Wallcur® PRACTI-Code Meds
Supplier: Wallcur
PRACTI-Code Meds
1510CP 1508AR 1514SF 1507N 1505E 1522LX 1518L 1504D 113HF 1519B 1506L 1502B 115TC 1500CP TCCC-100 1501A 1509AY 1503C
470232-730EA 10.39 CAD
470232-730 470354-928 470355-020 470355-030 470232-734 470232-722 470232-732 470232-742 470232-720 470354-986 470232-716 470232-726 470354-996 470232-714 470232-724 470232-718 470354-990 470232-728
Wallcur® PRACTI-Code Meds
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Preloaded Syringes For Your EMS, ACLS Or Nursing Training
  • Contains No Medication
  • Luer Lock Syringes Labeled And Prefilled
  • Individually Packaged For Loading Into Practice Crash Carts Or Field Bags
Fill or Refill your training response kits with these commonly used code meds. Each syringe is preloaded to accurately simulate the experience of drug administration in a pressure simulation. The luer lock connection matches up perfectly with IV connections. Supplied in 5 ml, 10 ml or 50 ml syringes, each is individually packaged and labeled to simulate a real ER experience. Use these tools to train and test students for: Accurate reading and opening crash cart code boxes, Hand/eye coordination while engaging glass syringe with plastic barrel, Dosage calculation and measurement, Intravenous administration of PRACTI-Crash Code™ solutions using an aseptic needless system assessment and Charting the amount, route, time, and efficacy of the delivery.
Available individually or in packages. The 6 pack kit contains one syringe each of PRACTI-Atropine, PRACTI-Sodium Bicarb, PRACTI-Calcium Cl, PRACTI-Dextrose, PRACTI-Epinephrine, and PRACTI- Lidocaine. The 9 pack kit contains one each of all 9 syringes.
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use