Wallcur® PRACTI-Ampules
Supplier: WALLCUR, LLC.
401PA 412MA 413MT 428AD 429PL 430PA 431PA 432PA 455CA 351AD 353MP 358HP 310PA
470232-796BX 167.89 CAD
470232-796 470232-816 470232-818 470232-848 470232-850 470232-852 470232-854 470232-856 470232-900 470358-280 470358-282 470358-284 470358-278
Wallcur® PRACTI-Ampules
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Prefilled And Etched For Proper Breakage.
  • Contains No Medication
  • Contains Distilled Water, Suitable For Use In Simulators And Injection Pads
  • Available Unlabeled, Or Labeled
  • Amber And Clear Versions Available To Simulate Light Sensitive Medications
Your students will learn how to accurately open, calculate, aspirate, and measure small and often hard to see solutions. Wallcur ampules are chemically scored for ease of use and to accurately simulate those found in hospitals and clinics. Unlabeled ampules can be combined with the PRACTI-Dose labels forMore Product Information