White Polypropylene Screw Caps, Pulp/Vinyl Liner,Qorpak®
Supplier: Qorpak
271085 263419 271086 270989 263420 263418 263421 263417 270984 270987 270997 270991 270986 270990
76012-880CS 210.11 CAD
76012-880 76012-860 76012-882 76012-870 76012-862 76012-884 76012-872 76012-864 76012-886 76012-874 76012-866 76012-876 76012-868 76012-878
White Polypropylene Screw Caps, Pulp/Vinyl Liner,Qorpak®
Closures Bottle Closures
Polypropylene caps are known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability.
  • Perfect for general use
  • Not recommended for active hydrocarbons or bleaches
Pulp/Vinyl Lined Polypropylene Caps are commonly used for food, beverage, medical and chemical applications packed at less than 120 °F., including mild acids, alkalis, solvents, alcohols, oils, and aqueous products.