Straight Sided Rounds, Amber, Wide Mouth, Qorpak®
Supplier: Qorpak
260300 262559 265844 271835 270408 270402 270219 271837 270409 270403 270229
75878-402CS 45.63 CAD
75878-402 75878-404 75878-406 75878-496 75878-498 75878-500 75878-502 75879-226 75879-228 75879-230 75879-232
Straight Sided Rounds, Amber, Wide Mouth, Qorpak®
Bottles Glass Bottles
Amber Straight Sided Round Glass Jars are designed without shoulders for maximum storage capacity and UV protection.
  • Minimal chemical interference with contents
  • Amber glass jars absorb all wavelengths in the ultraviolet spectrum
  • Ensure integrity for long term storage
  • Offer excellent protection for UV-sensitive contents
Also known as Paragon Jars, these multi-purpose jars are very versatile.