HyFlex® 11-939 Cut Resistant and Oil Repellent Gloves, Fully Coated, Ansell
Supplier: Ansell Healthcare
11939060 11939080 11939090 11939100 11939110
75927-850CS 5103.72 CAD
75927-850 75927-852 75927-854 75927-856 75927-858 75927-860
HyFlex® 11-939 Cut Resistant and Oil Repellent Gloves, Fully Coated, Ansell
Gloves
The lightest weight cut resistant and oil repellent glove providing enhanced worker comfort and safety, with strategically reinforced 'high wear' regions for extended-use life. These gloves are made up of Dyneema® diamond technology material.
  • 18 gauge liner featuring Dyneema® Diamond Technology provides high dexterity and comfort
  • Fully coated dip provides liquid repellence
  • Silicone free design insures full product protection
The HyFlex® 11-939 is an ultra-lightweight, cut resistant and oil repellent fully coated glove with reinforced thumb crotch that provides up to 12× the durability for extended wear.
Designed to feature a full range of Ansell Product Technologies designed for maximum worker and product protection. RIPEL™ Technology prevents liquids and oil from making skin contact keeping hands safe and dry. ANSELL GRIP™ Technology delivers superior grip performance for safer handling of wet or oily parts. FORTIX™ Technology provides extended glove life and up to high abrasion resistance. ZONZ™ Comfort Fit Technology provides dexterity and comfort.
The nitrile coating and integrated thumb crotch reinforcement (TCR) offer long-lasting protection and 12× better wear life than unreinforced gloves based on field tests.
Certifications: Meets ANSI Cut Level A2 and ANSI Abrasion Level 4.