Somso® Nursing Care Manikins
Supplier: MARCUS SOMMER
CLA1 CLA2
470233-236EA 8904.8 CAD
470233-236 470233-238
Somso® Nursing Care Manikins
Models Human Biology Models
For more advanced patient care practice!
  • A great starter manikin
  • Expandable over time
Standard nursing doll in natural size for nursing care comprising of the following parts: Removable head, removable eyes, dentures and tracheostomy, thorax with chest wall and moving arms with infusion and injection pads, lower body with abdominal wall and anus praeter abdominal wall, injection pads in the buttocks, movable mounted legs with injection pads in the thigh, lung alveolus, stomach, intestines, detachable female and male genitalia with rectum and bladder.
The advanced version includes lungs exhibiting closure, additional male genetalia and a higher capacity bladder (2 L).