COD Digestion vials, VWR Chemicals BDH®
Supplier: VWR International
CABDH0400-150
CABDH0400-25
CABDH0401-150
CABDH0401-25
CABDH0402-150
CABDH0402-25
CABDH0403-150
CABDH0403-25
CABDH0400-150PK
426.05
CAD
CABDH0400-150
CABDH0400-25
CABDH0401-150
CABDH0401-25
CABDH0402-150
CABDH0402-25
CABDH0403-150
CABDH0403-25
COD Digestion vials, VWR Chemicals BDH®
Environmental Test Kits
BDH COD Vials used in determination of Chemical Oxygen Demand (COD)
- Easy to Use
- Mercuric formulation follows EPA approved method
- Low Range, High Range, and Mercury Free High Range
Vials contain premeasured digestion reagents suited to your expected range of COD, so there is no measuring of harmful chemicals- simply add your sample to the vial and digest.
Caution: The vials contain hazardous chemicals, typically corrosive, acute toxic and irritant. GHS signal word: Danger