ActivArmr® Workguard™ 43-216 Leather Welding Gloves, Ansell
Supplier: Ansell Healthcare
43216090 43216100 43216110
75796-494CS 1639.56 CAD
75796-494 75796-496 75796-498
ActivArmr® Workguard™ 43-216 Leather Welding Gloves, Ansell
Gloves
Heavy-duty welding glove for demanding welding and thermal job tasks.
  • Provides EN 12477 Type-A protection
  • Suitable for stick and MIG welding, as well as torch cutting and grinding
  • Comfortable cotton lining provides thermal insulation and extended comfort
  • Long cuff extends protection over the forearm
ActivArmr® 43-216 has an improved three-dimensional thumb design that greatly increases fine dexterity for precise welding control. Its premium split cowhide leather and welted seams with Kevlar® thread ensure durability in use while providing comfort and protection for the wearer.
High levels of durability, control, and protection from heat, flame, sparks, and puncture.