HyFlex® 11-931 Cut Resistant and Oil Repellent Gloves, Palm Coated, Ansell
Supplier: Ansell Healthcare
11931090 11931100 11931110
75927-834CS 1370.02 CAD
75927-834 75927-836 75927-986 75927-988 75927-990 75927-992
HyFlex® 11-931 Cut Resistant and Oil Repellent Gloves, Palm Coated, Ansell
Gloves
The lightest weight cut resistant and oil repellent glove providing enhanced worker comfort and safety, with strategically reinforced 'high wear' regions for extended-use life. These gloves are made up of Dyneema® diamond technology material.
  • RIPEL™ Oil repellent technology for maintaining dry hands in oily environments
  • Thumb Crotch Reinforcement Technology for up to 12x the durability of the inherently weak thumb crotch region*
  • ANSELL GRIP™ Technology provides an average of 13% better grip performance for safe handling of Dry and Oily items**
The HyFlex® 11-931 is an ultra-lightweight, cut resistant and oil repellent palm coated glove with reinforced thumb crotch that provides up to 12x the durability for extended wear.
Designed to feature a full range of Ansell Product Technologies designed for maximum worker and product protection. RIPEL™ Technology prevents liquids and oil from making skin contact keeping hands safe and dry. ANSELL GRIP™ Technology delivers superior grip performance for safer handling of wet or oily parts. FORTIX™ Technology provides extended glove life and up to high abrasion resistance. ZONZ™ Comfort Fit Technology provides dexterity and comfort.
*compared to unreinforced TCR competitor gloves
**compared to competitive set 2017
18 gauge liner featuring Dyneema® Diamond Technology provides high dexterity and comfort. The nitrile coating and integrated thumb crotch reinforcement (TCR) offer long-lasting protection and 12x better wear life than unreinforced gloves based on field tests. Palm coated dip provides liquid repellence. A silicone free design insures full product protection.
Certifications: Meets ANSI Cut Level A2 and ANSI Abrasion Level 4.