ZING Green Safety Eco Safety Sign CAUTION Hearing Protection Required While Equipment is Operating
Supplier: ZING Enterprises
10126 10126A 10126S 20126 20126A 20126S
76185-514EA 39.41 CAD
76185-514 76185-516 76185-626 76185-996 76185-998 76186-000
ZING Green Safety Eco Safety Sign CAUTION Hearing Protection Required While Equipment is Operating
Signs
The sign is available in two sizes, 7 × 10", and 10 × 14", and three eco-friendly formats.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.