ZING Green Safety Eco Safety Sign WARNING Biological Hazard, ZING Enterprises
Supplier: ZING Enterprises
10059 10059A 10059S 20059 20059A 20059S
76185-720EA 38.99 CAD
76185-720 76185-722 76185-724 76185-726 76185-728 76185-730
ZING Green Safety Eco Safety Sign WARNING Biological Hazard, ZING Enterprises
Signs
The sign is available in 2 sizes, 7 x 10", and 10 x 14", and three eco-friendly formats.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.