ZING Green Safety Eco Safety Sign DANGER, Hazardous Chemicals
Supplier: ZING Enterprises
2982 2982A 2982S 1982 1982A 1982S
76184-962EA 46.56 CAD
76184-962 76184-964 76184-966 76185-148 76185-150 76185-152
ZING Green Safety Eco Safety Sign DANGER, Hazardous Chemicals
Signs
The sign is available in 2 sizes, 7 x 10", and 10 x 14", and three eco-friendly formats.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.