ZING Green Safety Eco Safety Sign CAUTION Radioactive Waste Prohibited Down Drain
Supplier: ZING Enterprises
10071 10071A 10071S 20071 20071A 20071S
76185-258EA 38.99 CAD
76185-258 76185-260 76185-262 76185-264 76185-266 76185-268
ZING Green Safety Eco Safety Sign CAUTION Radioactive Waste Prohibited Down Drain
Signs
The sign is available in two sizes, 7 × 10", and 10 × 14", and three eco-friendly formats.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.