ZING Green Safety Eco Safety Sign CAUTION X-Ray In Use
Supplier: ZING Enterprises
10070 10070A 10070S 20070 20070A 20070S
76185-246EA 38.99 CAD
76185-246 76185-248 76185-250 76185-252 76185-254 76185-256
ZING Green Safety Eco Safety Sign CAUTION X-Ray In Use
Signs
The sign is available in two sizes, 7 × 10", and 10 × 14", and three eco-friendly formats.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.