DAVISIL® Bonded Chromatographic Silica, W.R. Grace
Supplier: W.R. GRACE & CO. - CONN
DAVISIL® Bonded Chromatographic silica is an economical resin for a wide variety of bonding types, scale, and applications.
- Reversed and normal phase
- Scalable analytical to process
- Consistent and reliable performance
Our superior technology and scale-up capabilities make Grace the ideal partner for pharmaceutical and nutraceutical companies throughout the product development and commercial manufacturing process. Grace has a broad portfolio with the ability to scale to commercial scale volumes complimented by extensive experience in bonding different resins.