DAVISIL® Wide Pore Chromatographic Silica, W.R. Grace
Supplier: W.R. GRACE & CO. - CONN
5134301 5153368 5030057 5034754 5143586 5143590 5143587
76183-122EA 8780.68 CAD
76183-122 76183-126 76183-132 76183-134 76183-136 76183-138 76210-884
DAVISIL® Wide Pore Chromatographic Silica, W.R. Grace
Chromatography Sorbents HPLC Sorbents
Unique Wide Pore and Extra-Wide Pore silicas are a cost effective solution for purification of large molecules.
  • Pore Sizes 250 – 2500 Å
  • Exceptional mechanical strength
  • Large pore sizes with narrow pore size distribution
Grace Davisil® Wide Pore Chromatographic Silica is suitable for use as a packing for liquid chromatography normal phase separations which require pore sizes from 250 – 4500 Å. Applications include separation and purification of Biomolecules such as large peptides, oligonucleotides, proteins, vaccines, virus-like particles, RNA and DNA.