DAVISIL® 150Å Chromatographic Silica, W.R. Grace
Supplier: W.R. GRACE & CO. - CONN
5057993 5076059 5037727 5183331 5183333 5173139
76183-118EA 5176.16 CAD
76183-118 76183-120 76183-128 76183-178 76183-180 76210-928 76210-930
DAVISIL® 150Å Chromatographic Silica, W.R. Grace
Chromatography Sorbents HPLC Sorbents
Proven performance and high quality chromatographic silica.
  • Proven performance
  • Predicable scale-up
  • Exceptional quality and consistency
  • Wide selection
Grace Davisil® 150 Å Chromatographic Silica is suitable for use as a packing for liquid chromatography normal phase separations. Applications include separation and purification of small molecule pharmaceuticals, vitamins, natural products, and small peptides. Scalable from analytical to process scale.