DAVISIL® 60Å Chromatographic Silica, W.R. Grace
Supplier: W.R. GRACE & CO. - CONN
5022298
5029213
5037849
5055349
5136220
5153530
5183336
5183335
5183334
5183332
5054993
5191781
5191783
76183-110EA
20093.22
CAD
76183-110
76183-112
76183-114
76183-116
76183-124
76183-130
76183-170
76183-172
76183-174
76183-176
76210-922
76210-924
76210-926
DAVISIL® 60Å Chromatographic Silica, W.R. Grace
Chromatography Sorbents
HPLC Sorbents
Proven performance and high quality chromatographic silica.
- Proven performance
- Predicable scale-up
- Exceptional quality and consistency
- Wide selection
DAVISIL® 60 Å chromatographic silica is suitable for use as a packing for liquid chromatography normal phase separations. Applications include separation and purification of small molecule pharmaceuticals, organic synthesis and lipids. Scalable from analytical to process scale.