DAVISIL® Grade 923 Chromatographic Silica, W.R. Grace
Supplier: W.R. GRACE & CO. - CONN
5007039 5173172 5173173 5173174 5183337 5183338
76183-102EA 30459.19 CAD
76183-102 76183-104 76183-106 76183-108 76183-166 76183-168
DAVISIL® Grade 923 Chromatographic Silica, W.R. Grace
Chromatography Sorbents HPLC Sorbents
Grade 923 meets the specification for Silica in the ASTM D-1319 FIA test and the ASTM D-2549 Test.
  • Original silica specified in ASTM
  • Proven performance
  • Exceptional quality and consistency
Grace® Grade 923 Chromatographic Silica is a narrow pore type silica gel which meets the requirements for Silica Gel set forth in the procedure ASTM D 1319 and IP 156, as well as ASTM D 2549. Available in several convenient package configurations.
Certifications: Meets Specification for Silica in ASTM D-1319 and ASTM D-2549.