Omnifit® EZ SolventPlus™ Chromatography Columns with Two Adjustable Endpieces, Diba Industries
Supplier: Antyila Scientific
11943-98 11944-01 11944-04 11944-07 11944-10 11944-13 11944-16 11944-19 11944-22 11944-25 11944-28 11944-31 11944-34 11944-37 11944-40 11944-43 11944-46 11944-64 11944-67 11943-86 11943-89 11943-92 11943-95 11944-49 11944-52 11944-55 11944-58 11944-61 11944-70 11944-73 11944-76 11944-79
76175-868EA 1605.98 CAD
76175-868 76175-874 76175-880 76175-886 76175-892 76175-898 76175-904 76175-910 76175-916 76175-922 76175-928 76175-934 76175-940 76175-946 76175-952 76175-958 76175-964 76175-968 76175-974 76176-252 76176-258 76176-264 76176-270 76176-274 76176-280 76176-286 76176-292 76176-298 76176-332 76176-338 76176-344 76176-350
Omnifit® EZ SolventPlus™ Chromatography Columns with Two Adjustable Endpieces, Diba Industries
Chromatography Columns
PTFE, borosilicate glass, and Chemraz® wetted materials offer exceptional chemical resistance.
  • PEEK™ column threads offer outstanding durability
  • Ergonomically designed adjustable endpiece offers quick and easy height adjustment
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
Delivery information: Delivered with two (0.5 and 2 m length) ¹/₁₆" Ext.Ø PTFE tubing assembly with 1/4 - 28 UNF ETFE fitting at one end, a frit kit (contains two each of 10 µm PE, 10 µm PTFE, and 30 µm PTFE frits) and a Click-N-Seal® 10 to 32 PEEK conical port fitting.