Omnifit® EZ SolventPlus™ Chromatography Columns with One Fixed and One Adjustable Endpieces, Diba Industries
Supplier: Diba Industries
Omnifit®
11944-65 11944-71 11944-50 11944-32 11944-47 11944-23 11943-93 11944-11 11944-68 11944-53 11944-56 11944-29 11944-62 11943-90 11944-74 11944-44 11944-17 11944-77 11944-02 11944-14 11944-20 11944-41 11943-99 11944-05 11944-38 11944-80 11944-26 11944-59 11944-35 11944-08 11943-87 11943-96
76175-936EA 1832.95 CAD
76175-936 76175-912 76175-954 76175-876 76175-930 76175-894 76175-970 76175-870 76176-328 76176-346 76176-300 76176-266 76176-288 76176-282 76176-260 76175-906 76176-340 76175-948 76175-924 76175-966 76175-900 76175-888 76175-942 76175-864 76175-960 76175-882 76176-334 76176-276 76176-254 76176-352 76175-918 76176-294
Omnifit® EZ SolventPlus™ Chromatography Columns with One Fixed and One Adjustable Endpieces, Diba Industries
Chromatography Columns
PTFE, borosilicate glass, and Chemraz® wetted materials offer exceptional chemical resistance.
  • PEEK™ column threads offer outstanding durability
  • Ergonomically designed adjustable endpiece offers quick and easy height adjustment
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
Delivery information: Delivered with two (0.5 and 2 m length) ¹/₁₆" Ext.Ø PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end, a frit kit (contains two each of 10 µm PE, 10 µm PTFE, and 30 µm PTFE frits) and a Click-N-Seal® 10 to 32 PEEK conical port fitting.