CE UltraSpeed Pro™ Double Bucket System, Vileda Professional®
Supplier: Vileda Professional - FHP
157775 157777 157779
76170-718CS 1203 CAD
76170-718 76170-720 76170-722
CE UltraSpeed Pro™ Double Bucket System, Vileda Professional®
Mops
The easiest, most effective way to clean floors, walls, and ceilings within controlled environments.
  • Lightweight double bucket system allows for separation of dirty and clean liquid, minimizing cross contamination
  • Touchless wringing of mops
  • Compact chassis, minimal storage space required
  • Completely autoclavable with all stainless steel parts
  • Highly durable and withstands all common disinfectants
  • Disposable microfiber mops are pre-laundered
Demands on effectiveness, cost, ease of use, and staff training is constantly increasing
Designed for use in cleanrooms up to ISO 4-9/GMP A/B, C/D within Pharma, MedDevice, BioPharma, Healthcare, Microelectronics, and Semiconductor industries. CE UltraSpeed Pro™ outperforms current bucket and wringer systems in every aspect. All system parts are fully autoclavable. The Polyester Microfiber mops offer low particle shedding and outstanding cleaning performance. Touchless wringing of mops greatly reduces the risk of cross-contamination, and the user controls how wet the mop is for disinfection and removal of contamination. Additionally, the compact chassis is ideal for sites with limited storage. It is lightweight, ergonomic, and easy to use.
The CE UltraSpeed Pro™ mopping system has options to suit any site’s needs. The system can be used from small to large areas with optional ergonomic push bar attachment available. Disposable mops are made of 100% Polyester in a Polyester Microfiber blend. They are pre-laundered and offer an intensive cleaning performance due to the embossed surface pattern. Mops are easy gliding and have low particle shedding in use.
Whatever the cleaning requirements, CE UltraSpeed Pro™ will dramatically increase speed and cleaning performance, and improve versatility and ergonomics while decreasing total costs.
Ordering information: The CE UltraSpeed Pro™ Double Bucket System includes two buckets with flat mop wringer and chassis. The CE UltraSpeed Pro™ Frame is designed for use with handle #119966.