Tweezers Essence, WA1 Handle with 2A-BUIS Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
TIP-2A-BUI 10004-B2A-SM
103302-414EA 93.2 CAD
103302-414 103303-318
Tweezers Essence, WA1 Handle with 2A-BUIS Tips, Electron Microscopy Sciences
Forceps
These precision tweezers from Dumont made of high quality steels with perfect finish suits all application needs.
These tweezers have an anti-magnetic WA1 handle with replaceble 2A-BUIS tips made of boxwood (Buxus). Tips are attached with screws. Torque: cN.m 9 ±1. Length is 130mm.
Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.