Multi-Well Embedding Mold, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70914 70912 70913
103302-332EA 51.81 CAD
103302-332 103302-476 103302-482
Multi-Well Embedding Mold, Electron Microscopy Sciences
Moulds Moulds for Histology
Clear, white or blue silicone mold for embedding.
This unique silicone mold has 2 different cavity sizes to meet all embedding needs. 2 troughs: 1 L x 1/8 W x 1/8" D. 14 cavities: 1/4 L x 1/2 W x 1/8" D. 1 trough: 23/4 L x 3/32 W x 1/8" D. 10 cavities: 3/8 L x 3/4 W x 1/8" D. Each cavity has an alpha-numeric marking, A1-A7, B1-B7, C1-C5, D1-D5. Overall mold measures: 43/4 L x 3 W x 1/4" H (121 x 76 x 6 mm).