Immuno Mount with Para Phenylenediamine, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

17989-80 17989-81
103302-168EA 206.57 CAD
103302-168 103306-378
Immuno Mount with Para Phenylenediamine, Electron Microscopy Sciences
Stains Microscopy and staining reagents

EMS Immuno Mount™ with strong anti-fading agent, 1, 4-phenylenediamine (PPD) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.


  • Ready to use mounting medium with dark coffee color and this color does not interfere with Immunofluorescence
  • Refractive Index in solution: 1 400 ± 0.002


The unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Alexa fluoro 488, Alexa fluoro 594, tetramethyly rhodamine, and Redox. The fluorescence is retained during prolonged storage at 4 °C in the dark. Applications in immunofluorescence and confocal microscopy.
Storage: 2-8°C, DO NOT FREEZE.


Caution: Not suitable for immunofluorescence of Cy dyes, Phycoerythrin (R-PE), phyocyanin (PC), and allophycocyanin (APC)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR