Glycerol Fluoro Mount with Para Phenylenediamine, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
17989-70 17989-71
103302-166EA 214.26 CAD
103302-166 103306-376
Glycerol Fluoro Mount with Para Phenylenediamine, Electron Microscopy Sciences
Stains Microscopy and staining reagents
EMS Glycerol Fluorescence Mount with strong anti-fading agent, 1, 4-phenylenediamine (PPD) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.
  • Ready to use mounting medium with dark coffee color and this color does not interfere with Immunofluorescence
  • Refractive Index: 1.4563
The unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Alexa fluoro 488, Alexa fluoro 594, tetramethyly rhodamine, and Redox. The fluorescence is retained during prolonged storage at 4 °C in the dark. Application: Immunofluorescence, confocal microscopy. Storage: 2-8°C is recommended, Protect from light, DO NOT FREEZE
Caution: Not suitable for immunofluorescence of Cy dyes, Phycoerythrin (R-PE), phyocyanin (PC), and allophycocyanin (APC). DO NOT FREEZE.