Glycerol Mounting Medium with DABCO, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

17989-50 17989-51
103302-170EA 232.8 CAD
103302-170 103306-380
Glycerol Mounting Medium with DABCO, Electron Microscopy Sciences
Stains Microscopy and staining reagents

EMS Glycerol with anti-fading agent 1, 4-Diazobicyclo-(2,2,2-octane (DABCO™) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.


  • Ready to use
  • Refractive Index: 1.4615


The unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine, Redox. Fluorescence is retained during prolonged storage at 4°C in the dark.
Storage: 2-8°C is recommended, Protect from light


Caution: Not recommended for phycoethyrin (PE), phycocyanin (PC) and allophycocyanin (APC)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR