Forensic Gunshot Residue Field Kits, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
76480-13 76480-11 76480-15
103301-914EA 46.97 CAD
103301-914 103302-464 103302-484
Forensic Gunshot Residue Field Kits, Electron Microscopy Sciences
Specimen Mounting Supplies
Gun Shot Residue Kit for Scanning Electron Microscopy.
  • Powder-free nitrile gloves
  • Evidence label
  • Chain of custody label on box
  • Tamper-evident cardboard transport box (41/2 W x 33/4 D x 3" H)
  • Tamper evident seal
The certified SEM pin stubs have very low amounts of Pb, Sb and Ba, not detectable by SEM/EDX. The caps securely hold the GSR sample disc for easy GSR collection. Pre-labeled discs and tubes are both ready-for-sampling. 3-Disc GSR Field Kit has two sampling discs plus a control disc to enhance certainty of the analysis. 4-Disc GSR Field Kit is useful for sampling palm and back separately on both hands. 5-Disc GSR Field Kit is compatible with FBI requirements, with four discs plus a control disc.
Delivery information: Kit includes certified SEM pin stubs with double-coated adhesive carbon tabs in pre-labelled plastic tubes. A sturdy, sealed tamper evident transport box is also included.