Thumb Dressing Forceps, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
77937-23 77937-24
103063-086EA 97.08 CAD
103063-086 76375-982 76375-984
Thumb Dressing Forceps, Electron Microscopy Sciences
Forceps
These conventional forceps are very popular and are available in a variety of length, tip, and handle styles.
  • Made of premium grade, hardened stainless steel
  • Feature a protective coating for extra durability and protection from rust and corrosion
  • Satin-matte finish reduces eye-fatiguing glare