Athene Thin Bar with Center Mark Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
AT300-CU AT400-CU AT200-NI AT300-NI AT200-CU
103301-982EA 235.14 CAD
103301-982 103301-996 103302-158 103302-216 103302-686
Athene Thin Bar with Center Mark Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
These athene grids are used in transmission electron microscope.
  • Copper and nickel grids
  • Good handling characteristics
  • Individually inspected before being packed
Athene grids are renowned for the highest quality standards, exceptionally refined grid bars.