AirChek Badges, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64472-10 64472-20 64472-30 64472-40
103306-268EA 1295.77 CAD
103306-268 103306-396 103306-398 103306-400
AirChek Badges, Electron Microscopy Sciences
Gas Detection Systems
Monitor vapor levels, simply and effectively.
  • Clips onto technicians clothing
  • Record of name, location, time
Airchek Badges provide an innovative, simple and effective system for monitoring vapor levels, including Formaldehyde, Glutaraldehyde, Xylene, Toluene, and Isopropanol. The badges provide continuous sampling for personal monitoring and allowing the measurements required under Government regulations. One badge provides continuous sampling over 8 hours. A second badge can be used to supply information for analysis of 15-minute peak exposure periods.
Order Now
SPECIFICATIONS
Environmentally Preferable