WHEATON® MicroLiter Crimp-Top µLVials™, Component Kits, 11 mm, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
11-1421A 11-1430A
97047-298PK 262.31 CAD
97047-298 97047-299 97047-300 97047-301 97047-302 97047-304 97047-306 97047-308 97047-310 97047-316 97047-317 97047-318 97047-319 97047-320 97047-322 97047-324 97047-326 97047-328
WHEATON® MicroLiter Crimp-Top µLVials™, Component Kits, 11 mm, DWK Life Sciences
Vials
MicroLiter vials are identical, clean, and dependable, and are certified to ensure optimal performance and reproducible results.
  • Unassembled kits
  • Multiple configurations available
  • Made from Type 1 borosilicate glass or polypropylene and meet strict tolerances
MicroLiter vials are cleaned and packaged in a certified Class 10000 clean room according to a designed process, developed to eliminate chromatographic errors associated with vials.
Critical physical dimensions are fully tested, as well as chemical cleanliness testing to ensure that trace levels of residues are not present.