Vydac 238MS C18, HPLC Columns
Supplier: Avantor
238MS5.310 238GK54MS/N
75798-088EA 2285.91 CAD
75798-088 75798-090 75798-092 75798-094 75798-096 75798-098 75798-100 75798-102 75798-712 75798-714 75798-716 75798-718 75798-720 75798-722 75798-724 75798-726 75798-728 75798-730 75798-732 75798-734 75799-108 75799-194 75799-196 75799-198 75799-200 75799-202 75799-204 75799-206 75830-670 75830-678 75830-790 75831-450 76398-144 76398-150
Vydac 238MS C18, HPLC Columns
Chromatography Columns
Vydac MS columns are a later development from grace for the reversed phase HPLC separation of biomolecules.
  • Five reversed-phase chemistries
  • Excellent peak shape with little or no TFA
  • High protein recoveries make scale-up easy
  • USP L1
Monomeric bonding offers increased peptide interaction and generally yields higher peak counts. Use for same applications as 218MS, but offers different C18 selectivity.
A proprietary surface treatment and bonding process give Vydac MS columns unique selectivity. A variety of reversed phases makes this product line suitable for the analysis of small peptides to large intact, undenatured proteins.
Vydac is a registered trademark of W.R.Grace and Co.