Vydac 214MS C4, HPLC Columns
Supplier: Avantor
214MS5205 214MS5405 214MS5.07515 214MS5.315 214MS5215 214MS5415 214MS5.325 214MS52 214MS54 214MS1022 214FSK1210MS 214GD52MS/N 214GD54MS/N 214GK52MS/N 214GK54MS/N
75798-120EA 2285.91 CAD
75798-120 75798-122 75798-124 75798-126 75798-128 75798-130 75798-132 75798-134 75798-136 75798-138 75798-140 75798-142 75798-144 75798-146 75798-148 75798-150 75798-152 75798-154 75798-156 75798-158 75798-160 75798-162 75798-164 75798-166 75798-168 75798-170 75798-172 75798-174 75798-176 75798-178 75798-180 75798-182 75798-184 75798-186 75798-188 75799-208 75812-978 75812-980 75812-982 75812-984 75812-986 75830-758 75830-762 75830-774 75830-778 76398-054 76398-058 76398-060
Vydac 214MS C4, HPLC Columns
Chromatography Columns
Vydac MS columns are a later development from Grace for the reversed phase HPLC separation of biomolecules.
  • Five reversed-phase chemistries
  • Excellent peak shape with little or no TFA
  • High protein recoveries make scale-up easy
  • USP L26
  • Stainless steel column hardware
Lower capacity than C18 or C8, suitable for hydrophobic proteins or when minimal organic solvent is desired. Ideal for biomolecules >10K MW, undenatured intact proteins, antibodies, oligonucleotides, human growth hormone.
A proprietary surface treatment and bonding process give VydacMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable