MOPS-Na ≥99%, white powder

Supplier: CHEM-IMPEX INTERNATIONAL, INC

Synonyms: 3-(N-Morpholino)propanesulphonic acid sodium salt, Sodium 3-morpholinopropane-1-sulphonate, Sodium 4-morpholin-1-ylpropylsulphonate, 3-(N-Morpholino)propanesulfonic acid sodium salt, Sodium 3-morpholinopropane-1-sulfonate, MOPS sodium salt, Sodium 4-morpholin-1-ylpropylsulfonate

00743-100G 00743-1KG
CH00743-100GEA 126.4 CAD
CH00743-100G CH00743-1KG
MOPS-Na ≥99%, white powder
MOPS-Na
Formula: C₇H₁₄NNaO₄S
MW: 231.25 g/mol
MDL Number: MFCD00064350
CAS Number: 71119-22-7

Order Now

Specification Test Results

Purity Limit ≥99% (Assay, Anhydrous Basis)
Appearance White Powder
Storage Temperature Store at Room Temperature
pH 10.0 – 12.0 (1 M solution in H₂O)
Loss on Drying ≤2%
pKa 7.0-7.4 @ 25°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR