BRAND® Four Sided Clear Disposable Cuvettes for Fluorescence Applications, BrandTech® Scientific
Supplier: Brandtech
759030 759035 759128
10853-038PK 54.83 CAD
10853-038 10853-040 10853-042 10853-044
BRAND® Four Sided Clear Disposable Cuvettes for Fluorescence Applications, BrandTech® Scientific
Cuvettes Spectrophotometer Cuvettes
Premium quality cuvettes, including a new UV-transparent plastic with ultra-low auto fluorescence.
  • Manufactured under cleanroom conditions
  • Polystyrene and UV-Transparent versions
BRAND® Fluorescence cuvettes are molded in one of the world's largest cleanrooms for life science plastics, without human contact for added cleanliness
It is used for routine, visible light applications, and for highest signal/noise ratios. They feature a 10 mm light path to fit all standard fluorimeters.
Cuvettes are mold-cavity sorted for the highest reproducibility.