45° Pin Mount and 90° Profile, Combination Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75950-07-A 75950-07-B
103304-086EA 267.42 CAD
103304-086 103304-380
45° Pin Mount and 90° Profile, Combination Holder, Electron Microscopy Sciences
Specimen Mounting Supplies
For use in Hitachi S-800, S-4000, S-4100, S-4200, S-4300, S-4500, S-4700 and S-3600N SEMs.
  • Aluminum or brass with stainless steel allen set screws
  • Holds SEM pin mounts
The scanning electron microscope holder is ideal for cross sections up to 6.35 mm (1/4") thick.
Delivery information: Includes allen wrench.