Vydac 219TP Diphenyl, HPLC Columns
Supplier: Avantor
219TP5210 219TP5215 219TP5415 219TP54 219GCC510 219TP510 219TP1010 219FSK1210 219GD52/N 219GK52/N
75798-494EA 2040.68 CAD
75798-494 75798-496 75798-498 75798-500 75798-502 75798-504 75798-506 75798-508 75798-510 75798-512 75798-514 75798-516 75798-518 75812-914 75812-916 75812-918 75812-920 75812-922 75813-054 75831-426 75831-428 75831-430 75831-434 75831-436 75831-438 76398-078
Vydac 219TP Diphenyl, HPLC Columns
Chromatography Columns
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
  • USP L11
Lowest capacity first generation diphenyl phase. Recommended for polypeptides with aromatic side chains, large hydrophobic proteins, membrane-spanning peptides, lipid peptides, fusion proteins from inclusion bodies.
The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
Vydac is a registered trademark of W.R.Grace and Co.