Stirrer Bearings, Macro, Glass, 10 mm, Chemglass
Supplier: Chemglass
CG-2070-01 CG-2070-02 CG-2070-05
80071-724EA 222.82 CAD
80071-724 80071-726 80071-732
Stirrer Bearings, Macro, Glass, 10 mm, Chemglass
Stirrers Stirring Tools
Precision bore bearing for use in moderate vacuum with a CG-2074 or CG-2075 ground 10 mm stirrer shaft.
Caution: NOTE: Not for use with CG-2078 or CG-2079 polished stirrer shafts.