Eyesaver® Bar Activated Eyewash Stations, Speakman®
Supplier: SPEAKMAN COMPANY SE
SE-505 SE-545
470204-058EA 1549.5 CAD
470204-058 470204-060
Eyesaver® Bar Activated Eyewash Stations, Speakman®
First Aid Supplies Eyewash Stations and Eyewash Bottles
Wall mounted rectangular eyewash bowl.
  • Stainless steel bowl and trigger bar activator
  • Volume control
  • 3.0 gpm at 30 psi
These eyewashes are available in wall-mount, and pedestal-mount styles. They offer twin aerated sprays for gentle, yet thorough rinsing of the eyes and face.
Certifications: Meet ANSI Z358.1 standard.
Delivery information: The units are supplied with a rectangular stainless steel bowl and panic bar activating system.
Order Now
SPECIFICATIONS
Environmentally Preferable