Tabletop Mounted Eyewash, Speakman®
Supplier: Speakman
SE-572
470190-910EA 1010.65 CAD
470190-910
Tabletop Mounted Eyewash, Speakman®
First Aid Supplies Eyewash Stations and Eyewash Bottles
Twin aerated sprayheads conveniently located on the counter.
  • Tabletop mounted
  • Stay-open valve
  • Paddle handle activation
  • Left- and right-handed option
Speakman's SE-572 eyewash is counter mounted and designed with two aerated plastic spray heads which have automatic flow control. This eyewash is activated by a paddle handle and swivels 90°. The paddle handle is made of stainless steel and the ball valve is chrome-plated and made of female brass.
Certifications: Meets ANSI Z358.1.