BioFit MVMT™ Critical Performance Seating, BioFit

Supplier: BioFit
MP-TH-T-AV126 MP-MH-T-AV126
76124-532EA 4182.45 CAD
76124-532 76124-534 76124-536 76124-538 76124-542
BioFit MVMT™ Critical Performance Seating, BioFit
Chairs
MVMT™ (Movement) ergonomic seating is changing the dynamic of the seated worker experience in today’s workplace, combining a forward-looking European perspective with the BioFit tradition of technological precision and proven durability.

  • Intuitive functionality including weight-activated control and synchronous seat and backrest
  • Engineered for task-specific range of motion and 24/7/365 use
  • Lumbar support and 3" adjustable backrest height
  • Aesthetic complements interior design trends, positively impacting corporate image
  • Optional easy wipe down to ISO Class 3
More Product Information
Order Now

SPECIFICATIONS

Model 76124-532 76124-534 76124-536 76124-538 76124-542
Environmentally Preferable
For use with Low benches/desks Medium height benches High benches Low benches/desks High benches
Seat/back color Black Black Black Black Black
Seat/back material Vinyl Vinyl Vinyl Vinyl Vinyl
Backrest dimensions W×H 17.5×19.5" W×H 17.5×19.5" W×H 17.5×19.5" W×H 17.5×19.5" W×H 17.5×19.5"

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR