Avantor® Alltima, Cyano HPLC Columns
Supplier: HICHROM LIMITED
81189 88180 88182 88189 88191 96084/N
75797-264EA 1385.93 CAD
75797-264 75797-266 75797-268 75797-272 75797-274 75797-276 75797-278 75797-280 75830-854 76398-440 76398-442
Avantor® Alltima, Cyano HPLC Columns
Chromatography Columns
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
  • Base deactivated silica phases
  • Stable bonding for long column lifetime
  • Multiple selectivity options
  • USP L10
General purpose cyano suitable for normal or reversed phase use. Suitable for rugged normal phase applications.