Avantor® Alltima C18-LL, HPLC Columns
Supplier: Avantor
88069 88099 96285/N
75798-842EA 1759.17 CAD
75798-842 75798-844 75798-848 75798-850 75830-368 75830-930
Avantor® Alltima C18-LL, HPLC Columns
Chromatography Columns
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
  • Base deactivated silica phases
  • Stable bonding for long column lifetime
  • Multiple selectivity options
  • USP L1
Lower carbon load than traditional Alltima C18. Reversed phase applications that require a less hydrophobic C18 phase.