Avantor® Alltima C18, HPLC Columns
Supplier: Avantor
43853 88353 88352 81143 43851 81412 81382 81383 81387 81388 50605 88888 88370 88371 81140 81142 88383 88889 88059 88052 88051 88053 88056 11543 88055 88057 88307 88063 81105 88334 88335 96680/N 5174010/N 96080/N 96080SP/N AL-10C18-300DE AL-10C18-300DF AL-3C18-100SP7 AL-3C18-250D AL-3C18-75D AL-5C18-10CP2 AL-5C18-150DE AL-5C18-150DF AL-5C18-250DF AL-5C18-300DE AL-5C18-300DF
75797-168EA 3635.09 CAD
75797-168 75797-172 75797-174 75797-176 75797-178 75797-180 75797-182 75797-184 75797-186 75797-190 75797-192 75797-194 75797-196 75797-198 75797-202 75797-204 75797-206 75797-210 75797-212 75797-214 75797-220 75797-222 75797-224 75797-226 75797-228 75797-230 75798-818 75798-820 75798-822 75798-824 75798-828 75798-830 75798-832 75798-834 75813-068 75813-070 75813-072 75813-074 75813-076 75813-078 75830-382 75830-384 75830-796 75830-844 75830-846 75831-456 75831-458 76398-348 76398-390 76398-392 76398-394 76398-396 76398-398 76398-400 76398-410 76398-412 76398-414 76398-416 76398-418 76398-420 76398-422 76398-424 76398-426 76398-428 76398-430
Avantor® Alltima C18, HPLC Columns
Chromatography Columns
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
  • Base deactivated silica phases
  • Stable bonding for long column lifetime
  • Multiple selectivity options
  • USP L1
Classic reversed phase retention and selectivity. High quality hydrophobic general purpose C18.