Avantor® Genesis, Cyano HPLC Columns
Supplier: HICHROM LIMITED
FM15965E FM25965E FM15965ER
75799-006EA 2875.54 CAD
75799-006 75799-008 75830-612 75830-614 75830-616 76398-906
Avantor® Genesis, Cyano HPLC Columns
Chromatography Columns
Avantor® Genesis phases are based on high purity metal-free spherical silica.
  • Good peak shape and reproducibility
  • Long column lifetime
  • pH stability 1 to 10
These are suitable for the analysis of a wide range of analytes.