SW® PowerForm® PF-95BK Black 5.6 mil Sustainable Nitrile Exam Gloves
Supplier: SW Safety Solutions
N716882 N716883 N716884 N716885 N716886
76051-106BX 27.68 CAD
76051-106 76051-108 76051-110 76051-112 76051-114
SW® PowerForm® PF-95BK Black 5.6 mil Sustainable Nitrile Exam Gloves
Gloves
These gloves protect hands and the environment, with a sustainable single-use design that combines performance, durability, and tactile sensitivity for critical tasks.
  • EcoTek® sustainable technology
  • Consistent thickness for greater dexterity and tactile sensitivity
  • Fully textured for excellent wet and dry grip
  • Silicone-free for critical environments
  • Clean processing technology reduces potential skin irritation
  • Optimal thickness for durability and tactile sensitivity
  • Tear-resistant beaded cuff eases donning
  • 1.5 AQL exceeds laboratory standards
EcoTek® gloves tested to ASTM D5526-12 standard test method for determining anaerobic biodegradation of plastic materials under accelerated landfill conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated. For additional information on the test reports, contact cs@swssglobal.com.
Certifications: WBENC Women Owned; NCMSDC Minority Business; Small Business Enterprise; CE 0493. PowerForm® is dermatalogically approved by the Skin Health Alliance (SHA), and meets stringent standards that ensure skin safety.
Ordering information: Glove sizes are as follows; S (6.5 to 7), M (7.5 to 8), L (8.5 to 9), XL (9.5 to 10), XXL (10.5 to 11).