Science Take-Out® Brittle Bones: A Density Problem
Supplier: Science Take-Out
STO-154U STO-154 STO-154-NOTG
470232-166EA 287.75 CAD
470232-166 470232-165 470232-164
Science Take-Out® Brittle Bones: A Density Problem
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
What is Osteoporosis?
  • Available as a single kit with material for one student or group
  • Unassembled 10 pack for additional savings
Three generations of a family learn how osteoporosis (low bone density) results in bones that are brittle and easily broken. Students will design and assemble models of bones to represent bone densities at different ages to determine their densities. This activity also helps students learn to interpret models, graphs, news articles, and posters to relate bone density, osteoporosis, and bone fracture risk.